missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lyo-ready SuperScript™ III Flash Reverse Transcriptase

Product Code. 31662844
Change view
Click to view available options
Quantity:
2 kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31662844 2 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31662844 Supplier Invitrogen™ Supplier No. A40006521
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lyo-ready SuperScript III Flash RT is suitable for one-step RT-qPCR applications, helping ensure extremely short RT step time along with improved specificity, reliability, and thermostability.

Invitrogen Lyo-ready SuperScript III Flash Reverse Transcriptase joins the SuperScript product family as a reverse transcriptase designed for one-step RT-qPCR and point-of-care assay development. It is a novel SuperScript III Reverse Transcriptase variant, engineered for enhanced speed, thermostability, and inhibitor tolerance. Lyo-ready SuperScript III Flash Reverse Transcriptase provides enhanced sensitivity as low as five copies per reaction.

Benefits of Lyo-ready SuperScript III Flash Reverse Transcriptase include:

  • Excellent choice for one-step RT-qPCR
  • Fast cDNA synthesis (~1 minute) reduces the time to results
  • Hot-start enables high-throughput applications, benchtop stability, and improved specificity of one-step RT-qPCR
  • Thermostability up to 70°C allows elevated reaction temperatures and cDNA synthesis from complex RNA targets
  • High inhibitor tolerance makes the system less dependent on RNA sample purity
  • Commercial rights and customization enable product usage for commercial purposes

Hot start

Lyo-ready SuperScript III Flash Reverse Transcriptase is inactive at ambient temperatures due to the hot start mechanism, which enables room temperature reaction setup and improved specificity and consistency of RT-qPCR results. When used in conjunction with hot start DNA polymerase, such as Lyo-ready Platinum™ II Taq DNA Polymerase, a fully assembled master mix can be stored at room temperature for up to 24 hours, enabling high-throughput applications and compatibility with automation and point-of-care assay development. Due to enhanced specificity, this pair of enzymes also enables multiplex RT-qPCR of up to five targets.

Lyo-ready

Lyo-ready SuperScript III Flash Reverse Transcriptase is a glycerol-free enzyme that ensures compatibility with microfluidics, air-drying, and lyophilization technologies. For a full lyo-ready one-step RT-qPCR solution, we recommend using SuperScript III Flash Reverse Transcriptase together with Lyo-ready Platinum II Taq DNA polymerase. To learn more about Lyo-ready Enzymes for molecular diagnostics development visit the page thermofisher.com/lyo-ready.

OEM reverse transcriptase

To learn more about OEM and raw materials for molecular diagnostics, customization capabilities, and commercial rights, visit the page thermofisher.com/mdx.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 60 U/μL
Content And Storage •Lyo-ready SuperScript™ III Flash Reverse Transcriptase, 2 kU
•Store at -20°C.
Format Stand-alone Enzyme
PCR Method 1-step RT-qPCR
Reaction Speed Fast
Optimal Reaction Temperature 60°C
Quantity 2 kU
Ribonuclease H Activity Reduced
Shipping Condition Dry Ice
For Use With (Application) Virus Detection
Hot Start Build-In Hot Start
Reaction Format Separate components
Reagent Type Reverse Transcription
Starting Material RNA
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.