missing translation for 'onlineSavingsMsg'
Learn More

Lys05, MedChemExpress

Product Code. 16471587
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16471587

Brand: Medchem Express HY12855A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lys05 (Lys01 trihydrochloride) is a novel lysosomal autophagy inhibitor with IC50 values of 3.6, 3.8, 6 and 7.9 μM for 1205Lu, c8161, LN229 and HT-29 cell line in the MTT assay.

Lys05 (Lys01 trihydrochloride) is a novel lysosomal autophagy inhibitor with IC50 values of 3.6, 3.8, 6 and 7.9 μM for 1205Lu, c8161, LN229 and HT-29 cell line in the MTT assay.

Specifications

Chemical Name or Material Lys05
CAS 1391426-24-6
Color Light Brown
Purity Grade Notes Research
Molecular Formula C23H26Cl5N5
Quantity 50 mg
Synonym Lys01 trihydrochloride
Solubility Information DMSO : 25 mg/mL (45.48 mM; ultrasonic and adjust pH to 2 with HCl) ∣H2O : 6.4 mg/mL (11.64 mM; Need ultrasonic)
SMILES CN(CCNC1=CC=NC2=C1C=CC(Cl)=C2)CCNC3=CC=NC4=CC(Cl)=CC=C43.[H]Cl.[H]Cl.[H]Cl
Molecular Weight (g/mol) 549.75
Formula Weight 549.75
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.