missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Lysing Matrix E, 96-tube Rack

Product Code. 17737524 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 Ea.
10 Pk.
Unit Size:
Each
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17737524 1 Ea. Each
15252870 10 Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17737524 Supplier MP Biomedicals™ Supplier No. 116984001
  • ONLINE EXCLUSIVE PRICE
    286.00 €
    270.90 € / Pack of 1
    Save 15.10 €
    5% Off
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Lysing Matrix E tubes contain 1.4 mm ceramic spheres, 0.1 mm silica spheres, and one 4 mm glass bead. Used to grind soil, sediment, water, feces, sludge, compost, rhizosphere. For use on FastPrep™ instruments or any bead beater.

  • Product Overview:
    • Lysing Matrix E, 96-Well Rack comes with 1.2 mL cluster-strip tube and cap assemblies.
    • Each tube is pre-filled with 1.4 ceramic spheres, 0.1 mm silica spheres, and 1 (one) 4 mm glass bead.
    • Will not bind nucleic acids.
  • Applications: Lysing Matrix E can also be effective in lysing mixed samples, such as microbe infected tissue, and can be used in RNA extraction from tumors and other difficult tissues.
  • Sample Type: Feces, Soil, Wastewater, Environmental Water, Sludge, Tumor
  • Sample Volume: ≤80 mg per prep
TRUSTED_SUSTAINABILITY

Specifications

Bead Type 1.4 mm Ceramic Spheres, 0.1 mm Silica Spheres, 4 mm Glass Beads
Sample Type Soil, Environmental, Feces
DNase Dnase-free
For Use With (Equipment) FastPrep-96
For Use With (Application) Nucleic Acid Isolation and Purification, Homogenization
Matrix Lysing Matrix E
No. of Reactions 96
Quantity 1 Ea.
RNase RNase-free
Sample Volume less than 80 mg per prep

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.