missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Lysing Matrix S, Bulk

Product Code. 15252830 Shop All MP Biomedicals Products
Change view
Click to view available options
For Use With (Equipment):
All FastPrep Adapters
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15252830 All FastPrep Adapters 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15252830 Supplier MP Biomedicals™ Supplier No. 116925000
  • ONLINE EXCLUSIVE PRICE
    366.00 €
    346.50 € / 1kg
    Save 19.50 €
    5% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lysing Matrix S is used to break hard/brittle samples. Specialty applications include RNA and protein extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing.

  • Contains six (6) 1/8 in. (3.175 mm) diameter stainless steel beads.
  • Use to break hard/brittle samples.
  • Uses include RNA and protein extraction from difficult samples where lysis by impaction only is preferable.
  • Specialty applications include RNA and protein extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing.
  • Store at Room Temperature (15 to 30°C).

Specifications

Diameter (Metric) 3.175 mm
Material Stainless Steel
For Use With (Equipment) All FastPrep Adapters
Packaging Quantity 1 kg
Sample Type Bone, Heart, Lung, Seeds, Skeletal Muscle, Spores, Food, Plants, Roots, Fixed Samples, Tough Tissues, All Samples

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.