missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lysis Buffer, 0.5% Tween 20, pH 8.0

Product Code. 15454455
Change view
Click to view available options
Quantity:
500mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15454455 500mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15454455 Supplier Thermo Scientific™ Supplier No. J61792.AP
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For isolation of His-tag proteins

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 500mL
Format Liquid
Product Type Cell Lysis Reagent
For Use With (Application) For isolation of His-tag modified proteins
pH 8
Content And Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.