missing translation for 'onlineSavingsMsg'
Learn More

Abcam Lysis Buffer IV

Product Code. 32112781 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1 x 100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32112781 1 x 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32112781 Supplier Abcam Supplier No. ab3239661x100mL
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lysis Buffer IV (ab323966) is part of the reagents, controls & accessories range. Abcam offers high-quality biological reagents and tools including antibodies, proteins, assays, cell lines and lysates.

Lysis Buffer IV (ab323966) is part of the reagents, controls & accessories range. Abcam offers high-quality biological reagents and tools including antibodies, proteins, assays, cell lines and lysates.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Quantity 1 x 100 mL
Regulatory Status RUO
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.