missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Lysis Buffer M

Product Code. 31937803
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31937803 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31937803 Supplier Macherey-Nagel™ Supplier No. 743210.250
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lysis Buffer for Direct PCR from Mice

  • For use in a variety of lab applications

Specifications

pH 8.5 to 9.5
Quantity 250 mL
Solution Type Buffer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.