missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lysis Buffer for MagJET Plasmid DNA Kit

Product Code. 13515230 Shop All Thermo Scientific Products
Click to view available options
Quantity:
25 mL
90 mL
Unit Size:
25mL
90mL
This item is not returnable. View return policy

Product Code. 13515230

Brand: Thermo Scientific™ R2592

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Thermo Scientific MagJET Plasmid DNA Kit is designed for fast and efficient purification of plasmid DNA from overnight cultures of E. coli cultures. The kit utilizes magnetic bead technology, enabling high yields and robust performance.

The Thermo Scientific MagJET Plasmid DNA Kit is designed for fast and efficient purification of plasmid DNA from overnight cultures of E. coli cultures. The kit utilizes magnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.

TRUSTED_SUSTAINABILITY

Specifications

Buffer Lysis Buffer
Format Kit
Product Line MagJET
Product Type Lysis Buffer
Quantity 90 mL
compliance-icons
Product Identifier
  • Lysis Buffer for MagJET Plasmid DNA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : Sodium lauryl sulphate

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.