missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Lysis Buffer ML

Product Code. 12763411
Change view
Click to view available options
Quantity:
30 mL
Unit Size:
30mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12763411 30 mL 30mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12763411 Supplier Macherey-Nagel™ Supplier No. 740973.30
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lysis Buffer ML to be used with NucleoSpin miRNA kits.

Specifications

CAS 593-84-0
Color Colorless
For Use With (Application) Nucleic Acid purification
pH 6.5 to 7.5
Physical Form Liquid
Quantity 30 mL
Formulation Guanidinium thiocyanate; 2-Mercaptoethanol; Water
Odor Sulfuric
compliance-icons
Product Identifier
  • Buffer ML (30 mL)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Supplemental information
  • MIXTURE LIST-Contain: guanidinium thiocyanate, 2-mercaptoethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.