missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lysis Solution for GeneJET Plasmid Miniprep Kit

Product Code. 10397979 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
70 mL
Unit Size:
70mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10397979 70 mL 70mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10397979 Supplier Thermo Scientific™ Supplier No. R1223
Focus on Regenerative Therapies
Use promo code 28027
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific GeneJET Lysis Solution is a component of the GeneJET Plasmid Miniprep Kit and may be purchased separately.

Thermo Scientific GeneJET Lysis Solution is a component of the GeneJET Plasmid Miniprep Kit (Cat. No. K0502, K0503) and may be purchased separately.

Thermo Scientific GeneJET Lysis Solution is a component of the GeneJET Plasmid Miniprep Kit (Cat. No. K0502, K0503) and may be purchased separately.

Related Products

  • Resuspension Solution for GeneJET Plasmid Miniprep Kit (Cat. No. R1213)
  • Neutralization Solution for GeneJET Plasmid Miniprep Kit (R1233)
  • Wash Solution for GeneJET Plasmid Miniprep Kit (R1243)
  • Binding Buffer for GeneJET Plasmid Miniprep Kit (R1331)
  • GeneJET NGS Cleanup Kit (K0851)
  • Binding Buffer for GeneJET NGS Cleanup Kit (R2811)
  • Prewash Buffer for GeneJET NGS Cleanup Kit (R2821)
  • Wash Buffer for GeneJET NGS Cleanup Kit (R2831)
  • GeneJET Plasmid Miniprep Kit (K0502)
  • RNase A (R1253)
  • Elution Buffer for GeneJET Plasmid Miniprep Kit (R1263)
TRUSTED_SUSTAINABILITY

Specifications

Buffer Lysis Buffer
Content And Storage Store at room temperature,-20°C
Description Lysis Solution
Product Line GeneJET
Product Type Lysis Solution
Quantity 70 mL
Form Liquid
compliance-icons
Product Identifier
  • Lysis Solution
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contain: Sodium lauryl sulphate, Sodium hydroxide

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.