missing translation for 'onlineSavingsMsg'
Learn More

Melford Lysozyme from Chicken Egg White

Product Code. 30018447
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018447 1 g 1g
30018449 5 g 5g
30018448 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018447 Supplier Melford Supplier No. L381001.0
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Purified from chicken egg white, 2x crystallized, dialyzed and supplied as a salt free lyophilized powder.

Purified from chicken egg white, 2x crystallized, dialyzed and supplied as a salt free lyophilized powder.

Dry powder stable for 3 - 5 years when stored dry at 2°C to 8°C.

Stock solution is stable up to 3 days at 4°C and up to 1 year at -20°C.

Specifications

CAS 12650-88-3
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.