missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ m-3M3FBS

Product Code. 18044282
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18044282 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18044282 Supplier R&D Systems™ Supplier No. 1941/10
  • ONLINE EXCLUSIVE PRICE
    214.85 €
    193.20 € / 10mg
    Save 21.65 €
    10% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Phospholipase C activator

Activator of phospholipase C (PLC). Stimulates superoxide generation, Ca2+ release and inositol phosphate formation in a variety of cell types. Also inhibits growth of the leukemic cell lines U937 and THP-1.

Negative Control also available.

TRUSTED_SUSTAINABILITY

Specifications

CAS 200933-14-8
Quantity 10 mg
Product Line Tocris
Molecular Formula C16H16F3NO2S
Formula Weight 343.36
Purity >99%
Molecular Weight (g/mol) 343.36
Chemical Name or Material m-3M3FBS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.