missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ m-Chlorophenylbiguanide hydrochloride

Product Code. 18055942
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18055942 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18055942 Supplier R&D Systems™ Supplier No. 0440/100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective 5-HT3 agonist

Prototypical potent and selective 5-HT3 receptor agonist.
TRUSTED_SUSTAINABILITY

Specifications

CAS 2113-05-5
Quantity 100 mg
Product Line Tocris
Molecular Formula C8H11Cl2N5
Formula Weight 248.11
Purity >98%
Molecular Weight (g/mol) 248.11
Chemical Name or Material m-Chlorophenylbiguanide hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.