missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals m-Cresol Purple, pure, indicator grade

Product Code. 10792461 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10792461 5 g Glass bottle 5g
10013030 10 g Glass bottle 10g
13418488 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10792461 Supplier Thermo Scientific Chemicals Supplier No. 228200050
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2303-01-7 | C21H18O5S | 382.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2303-01-7
Molecular Formula C21H18O5S
Molecular Weight (g/mol) 382.43
MDL Number MFCD00005871
InChI Key OLQIKGSZDTXODA-UHFFFAOYSA-N
PubChem CID 73030
IUPAC Name 4-[3-(4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
SMILES CC1=C(C=CC(=C1)O)C2(C3=CC=CC=C3S(=O)(=O)O2)C4=C(C=C(C=C4)O)C

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Specific Extinction 900 min. (578nm, in buffer)
Visual Transition Interval from pH 7.4 (yellow) to pH 9.0 (purple)
Color Green
Quantity 5 g
Physical Form Fine Crystalline Powder
Formula Weight 382.42
Grade Pure
Chemical Name or Material m-Cresol Purple

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.