missing translation for 'onlineSavingsMsg'
Learn More

m-Ethyltoluene, 98%

Product Code. 15294604 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15294604 10 g 10g
15204614 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15294604 Supplier Thermo Scientific Acros Supplier No. 119000100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 620-14-4 | C9H12 | 120.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-14-4
Molecular Formula C9H12
Molecular Weight (g/mol) 120.20
MDL Number MFCD00009259
InChI Key ZLCSFXXPPANWQY-UHFFFAOYSA-N
Synonym 3-ethyltoluene, m-ethyltoluene, 1-methyl-3-ethylbenzene, benzene, 1-ethyl-3-methyl, m-ethylmethylbenzene, m-methylethylbenzene, toluene, m-ethyl, 3-methylethylbenzene, 1-ethyl-3-methyl-benzene, unii-737ptd7o7e
PubChem CID 12100
ChEBI CHEBI:77512
IUPAC Name 1-ethyl-3-methylbenzene
SMILES CCC1=CC=CC(C)=C1

Specifications

Melting Point -97°C to 95°C
Density 0.8600g/mL
Boiling Point 158.0°C to 159.0°C
Flash Point 38°C
Assay Percent Range 97.5% min. (GC)
Linear Formula C2H5C6H4CH3
Refractive Index 1.4956
Quantity 10 g
Beilstein 05,396
Specific Gravity 0.86
Formula Weight 120.19
Percent Purity 98%
Chemical Name or Material m-Ethyltoluene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • m-Ethyltoluene, 98%
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.