missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ M.R.S. Agar

Product Code. p-200112471
Click to view available options
Quantity:
500 g
2.5 kg
Unit Size:
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10209632

Brand: Thermo Scientific™ CM0361B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate and enumerate lactic acid bacteria from food samples with this medium.

Isolate and enumerate lactic acid bacteria from food with Thermo Scientific™ Oxoid™ M.R.S. Agar (Dehydrated). This medium is designed to support profuse growth of all strains of lactobacilli, even the difficult and slow-growing strains of Lactobacillus brevis and Lactobacillus fermenti.

M.R.S. Agar is designed to encourage the growth of the ‘lactic acid bacteria’ which includes species of the following genera: Lactobacillus, Streptococcus, Pediococcus, and Leuconostoc. All these species can produce lactic acid in considerable amounts.

  • Superior performance over the tomato juice medium of Briggs1 and the meat extract tomato juice medium of de Man
  • Allows growth of growth of all strains of lactobacilli, especially the difficult and slow-growing strains of Lactobacillus brevis and Lactobacillus fermenti

‘Lactic acid bacteria’ are Gram-positive, catalase and oxidase negative and are fastidious in their nutritional requirements. Growth is enhanced considerably by microaerobic conditions. Generally, the ‘lactic acid bacteria’ show delayed growth and smaller colony size than other microorganisms. They may be overgrown in non-selective media, especially if incubation is required for 2-4 days.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AFNOR, IDF, NMKL standards.
Description M.R.S. Agar (Dehydrated)
Form Powder
Product Type Agar
Quantity 500 g
Yield For 8.1L medium
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.