missing translation for 'onlineSavingsMsg'
Learn More

m-Toluoyl chloride, 99%

Product Code. 11406776
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11406776 100 g 100g
11496766 25 g 25g
11416776 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11406776 Supplier Thermo Scientific Alfa Aesar Supplier No. B23914.22
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1711-06-4 | C8H7ClO | 154.593 g/mol

m-Toluoyl chloride has been used to derivatize the C1-C4 aliphatic amines for their determination in air by HPLC. It has been used in determination of atmospheric ammonia by denuder-sampling and HPLC-UV detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Toluoyl chloride has been used to derivatize the C1-C4 aliphatic amines for their determination in air by HPLC. It has been used in determination of atmospheric ammonia by denuder-sampling and HPLC-UV detection.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, active metals, amines and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1711-06-4
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.593
MDL Number MFCD00000681
InChI Key YHOYYHYBFSYOSQ-UHFFFAOYSA-N
Synonym m-toluoyl chloride, m-methylbenzoyl chloride, 3-toluoyl chloride, toluoyl chloride, benzoyl chloride, 3-methyl, m-toluyl chloride, meta methyl benzoyl chloride, 3-methylbenzoylchloride, 3-toluoylchloride, unii-w2vjw4350k
PubChem CID 74375
IUPAC Name 3-methylbenzoyl chloride
SMILES CC1=CC=CC(=C1)C(=O)Cl

Specifications

Melting Point -23°C
Density 1.173
Boiling Point 220°C
Flash Point 76°C (168°F)
Odor Pungent
Refractive Index 1.5485
Quantity 100 g
UN Number UN3265
Beilstein 878419
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 154.6
Percent Purity 99%
Chemical Name or Material m-Toluoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.