missing translation for 'onlineSavingsMsg'
Learn More

m-Toluoylacetonitrile, 97%

Product Code. 11443984
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11443984 5 g 5g
11433984 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11443984

Brand: Thermo Scientific Alfa Aesar A11965.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 53882-81-8 | C10H9NO | 159.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53882-81-8
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
MDL Number MFCD00067922
InChI Key IVLKDYOTZMFMLO-UHFFFAOYSA-N
Synonym 3-methylbenzoylacetonitrile, 3-3-methylphenyl-3-oxopropanenitrile, 3-toluoylacetonitrile, m-toluoylacetonitrile, 3-oxo-3-m-tolyl propanenitrile, 3-oxo-3-m-tolylpropanenitrile, pubchem12073, maybridge1_004502, acmc-1alb6, 3-m-tolyl-3-oxo-propanenitrile
PubChem CID 143105
IUPAC Name 3-(3-methylphenyl)-3-oxopropanenitrile
SMILES CC1=CC=CC(=C1)C(=O)CC#N

Specifications

Melting Point 71°C to 75°C
Quantity 5 g
Beilstein 2717335
Formula Weight 159.19
Percent Purity 97%
Chemical Name or Material m-Toluoylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.