missing translation for 'onlineSavingsMsg'
Learn More

m-Tolyl isocyanate, 99%

Product Code. 11348226
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348226 5 g 5g
11358226 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348226 Supplier Thermo Scientific Alfa Aesar Supplier No. L15255.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 621-29-4 | C8H7NO | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-29-4
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00002021
InChI Key CPPGZWWUPFWALU-UHFFFAOYSA-N
Synonym m-tolyl isocyanate, benzene, 1-isocyanato-3-methyl, meta-tolyl isocyanate, m-tolylisocyanate, m-isocyanatotoluene, 3-methylphenyl isocyanate, 3-tolyl isocyanate, 3-methylphenylisocyanate, 1-isocyanato-3-methyl-benzene, m-methylphenyl isocyanate
PubChem CID 69303
IUPAC Name 1-isocyanato-3-methylbenzene
SMILES CC1=CC=CC(=C1)N=C=O

Specifications

Density 1.033
Boiling Point 75°C to 76°C (12 mmHg)
Flash Point 65°C (149°F)
Odor Pungent
Refractive Index 1.5305
Quantity 5 g
UN Number UN2206
Beilstein 386149
Sensitivity Moisture sensitive
Formula Weight 133.15
Percent Purity 99%
Chemical Name or Material m-Tolyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.