missing translation for 'onlineSavingsMsg'
Learn More

m-Xylene, 99+%, extra pure

Product Code. 10592324 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
10 L
Packaging:
Glass Bottle
Plastic Drum
Unit Size:
1 L
10 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10592324 10 L Plastic Drum 10 L
10543741 250 mL Glass Bottle 250mL
10674682 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10592324 Supplier Thermo Scientific Acros Supplier No. 180860100
  • ONLINE EXCLUSIVE PRICE
    655.00 €
    635.00 € / 10 L
    Save 20.00 €
    3% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 108-38-3 | C8H10 | 106.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-38-3
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008536
InChI Key IVSZLXZYQVIEFR-UHFFFAOYSA-N
Synonym m-xylene, 1,3-dimethylbenzene, m-xylol, m-dimethylbenzene, meta-xylene, m-methyltoluene, 3-xylene, benzene, 1,3-dimethyl, 1,3-dimethylbenzol, santosol 150
PubChem CID 7929
ChEBI CHEBI:28488
IUPAC Name 1,3-xylene
SMILES CC1=CC(C)=CC=C1

Specifications

Melting Point -48.0°C
CAS Min % 99.0
Color Colorless
CAS Max % 100.0
Density 0.8640g/mL
Quantity 10 L
Flash Point 25°C
Infrared Spectrum Authentic
Packaging Plastic Drum
Physical Form Liquid
Refractive Index 1.4960 to 1.4980
Residue after Evaporation 0.005% max.
Assay Percent Range 99+%
Specific Gravity 0.864
Linear Formula C6H4(CH3)2
Viscosity 0.62 mPa.s (20°C)
Water 0.1% max.
Beilstein 05, 370
Merck Index 15, 10278
Boiling Point 139.0°C
Solubility Information Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, ether and most organic, solvents
Formula Weight 106.17
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material m-Xylene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.