Product Code. 10674682
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic Drum
Unit Size:
1 L
10 L
2.5 L
250mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
Unit Size
10674682
1 L
Glass Bottle
1 L
10543741
250 mL
Glass Bottle
250mL
10205570
2.5 L
Glass Bottle
2.5 L
10592324
10 L
Plastic Drum
10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code.
10674682
Supplier
Thermo Scientific Acros
Supplier No.
180860010
76.18 EUR valid until 2026-12-31 BEST PRICE on Promo! Use promo code "28038" to get your promotional price.
missing translation for 'validMainframeMsg'
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
CAS: 108-38-3 | C8H10 | 106.17 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
CAS
108-38-3
Molecular Formula
C8H10
Molecular Weight (g/mol)
106.17
MDL Number
MFCD00008536
InChI Key
IVSZLXZYQVIEFR-UHFFFAOYSA-N
Synonym
m-xylene, 1,3-dimethylbenzene, m-xylol, m-dimethylbenzene, meta-xylene, m-methyltoluene, 3-xylene, benzene, 1,3-dimethyl, 1,3-dimethylbenzol, santosol 150
PubChem CID
7929
ChEBI
CHEBI:28488
IUPAC Name
1,3-xylene
SMILES
CC1=CC(C)=CC=C1
Specifications
Melting Point
-48.0°C
CAS Min %
99.0
Color
Colorless
CAS Max %
100.0
Density
0.8640g/mL
Quantity
1 L
Flash Point
25°C
Infrared Spectrum
Authentic
Packaging
Glass Bottle
Physical Form
Liquid
Refractive Index
1.4960 to 1.4980
Residue after Evaporation
0.005% max.
Assay Percent Range
99+%
Specific Gravity
0.864
Linear Formula
C6 H4 (CH3 )2
Viscosity
0.62 mPa.s (20°C)
Water
0.1% max.
Beilstein
05, 370
Merck Index
15, 10278
Boiling Point
139.0°C
Solubility Information
Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, ether and most organic, solvents
Formula Weight
106.17
Percent Purity
≥99%
Grade
Extra Pure
Chemical Name or Material
m-Xylene
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?