missing translation for 'onlineSavingsMsg'
Learn More

m-Xylene, 99%

Product Code. 11336656
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11336656 2500 mL 2.5 L
11316656 100 mL 100mL
11326656 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 11336656 Supplier Thermo Scientific Alfa Aesar Supplier No. L03788.0F
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

CAS: 108-38-3 | C8H10 | 106.17 g/mol

m-Xylene is used in the production of isophthalic acid, which is a monomer and utilized in preparation of polyethylene terephthalate. It is used as an important starting material in the production of 2,4- and 2,6-xylidine. It is used as solvent in histology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Xylene is used in the production of isophthalic acid, which is a monomer and utilized in preparation of polyethylene terephthalate. It is used as an important starting material in the production of 2,4- and 2,6-xylidine. It is used as solvent in histology.

Solubility
Miscible with organic solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.  
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-38-3
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008536
InChI Key IVSZLXZYQVIEFR-UHFFFAOYSA-N
Synonym m-xylene, 1,3-dimethylbenzene, m-xylol, m-dimethylbenzene, meta-xylene, m-methyltoluene, 3-xylene, benzene, 1,3-dimethyl, 1,3-dimethylbenzol, santosol 150
PubChem CID 7929
ChEBI CHEBI:28488
IUPAC Name 1,3-xylene
SMILES CC1=CC(C)=CC=C1

Specifications

Melting Point -47°C
Density 0.866
Quantity 2500 mL
Flash Point 25°C (77°F)
Odor Mild
Refractive Index 1.497
UN Number UN1307
Beilstein 605441
Merck Index 14,10081
Boiling Point 138°C to 139°C
Solubility Information Miscible with organic solvents. Immiscible with water.
Formula Weight 106.17
Percent Purity 99%
Chemical Name or Material m-Xylene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • m-Xylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: m-Xylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.