missing translation for 'onlineSavingsMsg'
Learn More

Micronova M-Zone™ MegaSorb™ Microfiber Looped String Mop, 12 oz.

Product Code. 32179539 Shop All Micronova Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color
32179539 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32179539 Supplier Micronova Supplier No. MZCRMS1112IR
  • ONLINE EXCLUSIVE PRICE
    408.00 €
    376.67 € / Pack of 12
    Save 31.33 €
    7% Off
 Restricted Item
Estimated Shipment: 01-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A highly absorbent mop that combines the cleaning efficiency of microfiber with the ease of a looped string mop. The tufted strands are designed to pick up liquids and tiny particles in controlled and critical environments.

  • Microfiber looped string mop
  • Highly absorbent and low-linting
  • Polyester and Nylon tufted microfiber tubes
  • Works to pick up tiny particles where cotton fibers just roll over
  • Glides easily around cleanroom equipment and fixtures
  • Designed for use in controlled and critical environments
  • White microfiber option includes an additional scrubbing band
  • Use with MH-1 Handle or QDMH-1 MicroMop adapter with SSU series handles
  • Bulk packed option available
  • Compatible with alcohol and strong disinfectants
  • Launderable for reuse

Specifications

Product Type String Mop
Color White
Sterility Non-sterile
Autoclavable Autoclavable
Disposable Reusable
Color-coded No
Material Synthetic Microfiber
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.