Learn More
Micronova™ M-Zone™ Polyester Snap Mop
BIOPHARMAGREENER_CHOICE
Product Code. 15551755 Shop All Micronova Products
Change view
Click to view available options
Material:
Polyester
Unit Size:
Pack of 24
Product Code. Material Unit Size
15551755 Polyester Pack of 24
1 options
Product Code. 15551755 Supplier Micronova™ Supplier No. MZSM1114IR
  • ONLINE EXCLUSIVE PRICE
    544.00 €
    517.36 € / Pack of 24
    Save 26.64 €
    4% Off
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Polyester mop with waffle weave for increased absorbency. Micronova™ M-Zone™ Polyester Snap Mop is compatible with strong disinfectants.
  • Irradiated
  • Autoclavable
  • For use with QDSL and QPSL series mop adapters
  • Designed for clean and controlled environment
  • 100% Polyester with waffle weave for increased absorbency
  • Compatible with strong disinfectants
  • Autoclavable
Certifications
ISO 9001:2008
Compliance
AMI Guidelines for Sterilization
Specifications
Material Polyester