Learn More
Applied Biosystems™ M13 Human, Forward Primer, HPLC
Product Code. 15534374 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
50 nmol
Unit Size:
Each
Product Code. Quantity Unit Size
15534374 50 nmol Each
1 options
Product Code. 15534374 Supplier Applied Biosystems™ Supplier No. A15635
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
M13 Human, Forward Primer, HPLC
Use our tool to find and order pre-designed primers for PCR and Sanger sequencing. Free of SNPs and dimers, target-specific, and suitable for universal PCR conditions, these primers can be ordered unmodified, M13-tailed, HPLC-purified, or desalted. All primers are checked by mass spectrometry.
TRUSTED_SUSTAINABILITY
Specifications
Description M13 Human Forward HPLC Primer, 50 nmole, For PCR and Sanger Sequencing
Product Type M13 Human Forward HPLC Primer
Primer Forward
Quantity 50 nmol
Grade HPLC