missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ M13 Sequencing Reverse Primer, HPLC

Product Code. 15564374 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
50 nmol
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15564374 50 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15564374 Supplier Applied Biosystems™ Supplier No. A15638
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

M13 Sequencing Reverse Primer, HPLC

Use our tool to find and order pre-designed primers for PCR and Sanger sequencing. Free of SNPs and dimers, target-specific, and suitable for universal PCR conditions, these primers can be ordered unmodified, M13-tailed, HPLC-purified, or desalted. All primers are checked by mass spectrometry.

TRUSTED_SUSTAINABILITY

Specifications

Delivery Type Single Tube, Dried-down
For Use With (Equipment) Mass Spectrometry
Description M13 Reverse Sequencing Primer, 50 nmole, HPLC Purification, 900pmole Minimal Spec Yield, 5' CAGGAAACAGCTATGACC Sequence, Delivered in a Single tube, Dried-down, Used for BigDye™ Cycle Sequencing Primers
Product Type M13 Reverse Sequencing Primer
Protein Degradation Sequence 5' CAGGAAACAGCTATGACC
Primer M13 Reverse Sequencing
Quantity 50 nmol
Grade HPLC
For Use With (Application) BigDye™ Cycle Sequencing Primers
Form Solid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.