missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MAbPac™ Reversed Phase HPLC Columns

Use the new Thermo Scientific™ MAbPac™ RP columns for high-performance reversed-phase (RP) HPLC and LC/MS characterization of monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins.

Brand:  Thermo Scientific™ 088646

Code : NEW

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 15427162

  • 215.78 € / Pack of 2
Estimated Shipment: 10-05-2024
to see stock.
Add to basket

Description

Description

Use the new Thermo Scientific™ MAbPac™ Reversed Phase columns for high-performance reversed-phase (RP) HPLC and LC/MS characterization of monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins. These columns have excellent resolution, long column lifetime, and low carry-over. The surface chemistry, particle size, and pore size, have been optimized for high resolution separations.

Compatible with organic solvent used in HPLC and MS under RP conditions
  • Resin is based on a hydrophobic supermacroporous 4μm polymer particles
  • Stable at extreme pH (0 to 14) and high temperature (up to 110 °C)
Efficient with low carry over
  • 1,500Å large pore size polymeric resin
  • Efficient separation of protein molecules with very low carry over
Designed for reversed-phase biomolecule LC and LC/MS separations
  • Superior resolution power for monoclonal antibodies and related substances
  • High efficiency with low carry-over
  • Excellent MS compatibility
  • Wide operating pH range: 0 to 14
  • High temperature stability: up to 110°C
  • High throughput
Specifications

Specifications

Guard Column
10 mm
4 μm
1500 Å
Phenyl (Ph)
DVB (Divinylbenzene) Polymer
Reversed Phase
3 mm
4000 psi (275 bar)
0 to 14
<110°C
MAbPac
Reversed Phase
Bio-LC
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.