missing translation for 'onlineSavingsMsg'
Learn More

Cytiva MabSelect™ Chromatography Media

Product Code. 11339981 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 mL
200 mL
Unit Size:
200mL
25mL
Product Code. Quantity unitSize
11339981 25 mL 25mL
11359981 200 mL 200mL
2 options
This item is not returnable. View return policy

Product Code. 11339981

Brand: Cytiva 17519901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Protein A affinity medium designed for capturing MAbs from large sample volumes

  • High-flow agarose matrix allows for high flow velocities at production scale
  • BioProcess medium supported for industrial applications and well-established in approved processes
  • Low non-specific binding resulting in low levels of impurities in the product eluate pool
  • Oriented coupling of ligand and optimized matrix gives high dynamic binding capacity
  • Based on highly cross-linked agarose matrix with recombinant protein A ligand
  • Ligand is engineered to favor oriented coupling and expressed in E. coli and is free of components of mammalian origin
  • Hydrophilic nature of the base matrix ensures low levels of non-specific

  • Average particle size: 85μm (d50V)
  • pH Stability cleaning-in-place (CIP): 2 to 12
  • pH Stability working range: 3 to 10
  • Ligand: recombinant protein A (E.coli)
  • Matrix: highly cross-linked agarose
  • Binding capacity/mL chromatography medium: ∽30mg human lgG/mL at 2.4 min. residence time
  • Pressure/Flow specification: <500cm/hr., <200 kPa, BPG 300, bed height 20cm
TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mL
For Use With (Application) For Industrial AppliCatIons and Well-Established in Approved Processes
Storage Requirements 2°C to 8°C
Binding Capacity 30 mg Human lgG/mL (at 2.4 min. residence time)
Product Type MabSelect Media
Flow Rate <500 cm/Hoμr
Particle Size 85 μm
Height (English) Bed 7.87 in.
Height (Metric) Bed 20 cm
Includes Certificate of Analysis
Max. Pressure <200 kPa
Matrix Highly Cross-Linked Agarose
pH Range 3 to 10
Ligand Type Recombinant Protein A (E. coli)
Show More Show Less
compliance-icons
Product Identifier
  • MabSelect, 25 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.