missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MabSelect PrismA has been improved with an optimized high-flow agarose base matrix and a genetically engineered protein A-derived ligand, allowing future demands in mAb processing to be met.
Specifications
Specifications
| Matrix | Highly cross-linked agarose, spherical. |
| Content And Storage | 2 to 8°C, 20% ethanol |
| Max. Operating Pressure | 300 cm/hr. |
| Particle Size | 60 μm |
| Quantity | 25 mL |
| For Use With (Application) | Antibody affinity chromatography |
| Type | Chromatography Resin |
| Ligand Type | Alkaline-stabilized protein A-derived (E.coli). |
| Binding Capacity | 80 mg Human IgG/mL Resin at 6 min residence time. 65 mg Human IgG/mL Resin at 4 min residence time. |
Safety and Handling
Product Identifier
- MabSelect PrismA
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
- MIXTURE LIST-Contain: ethanol
- UNKNOWN ACUTE- per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?