missing translation for 'onlineSavingsMsg'
Learn More

Cytiva MabSelect PrismA Chromatography Resin

Product Code. 15883965 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 mL
200 mL
Unit Size:
200mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15883965 200 mL 200mL
15873965 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15883965 Supplier Cytiva Supplier No. 17549802
Powering Proteomics
Use promo code 28003
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Protein A Chromatography Resin

MabSelect PrismA has been improved with an optimized high-flow agarose base matrix and a genetically engineered protein A-derived ligand, allowing future demands in mAb processing to be met.

TRUSTED_SUSTAINABILITY

Specifications

Binding Capacity 80mg human IgG/mL resin at 6 min residence time. 65mg human IgG/mL resin at 4 min residence time.
Matrix Highly cross-linked agarose, spherical.
Content And Storage 2 to 8°C, 20% ethanol
Max. Operating Pressure 300 cm/hr.
Particle Size 60 μm
Quantity 200 mL
For Use With (Application) Antibody affinity chromatography
Type Chromatography Resin
Ligand Type Alkaline-stabilized protein A-derived (E.coli).
compliance-icons
Product Identifier
  • MabSelect PrismA
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE- per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.