missing translation for 'onlineSavingsMsg'
Learn More

Cytiva MabSelect Sure™ Chromatography Media

Product Code. 11392500 Shop All Cytiva Products
Click to view available options
Quantity:
200 mL
Unit Size:
200mL
This item is not returnable. View return policy

Product Code. 11392500

Brand: Cytiva 17543802

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MabSelect™ SuRe™ is a protein A affinity medium for capturing MAbs at process scale with an alkali-tolerant rProtein A ligand that allows the for cost-effective sanitization protocols

Has an alkali-tolerant rProteain A ligand designed to enhance protease resistance to produce monoclonal antibodies

  • Withstands rigorous CIP and sanitization procedures with 0.1 to 0.5 M NaOH
  • Design gives enhanced protease resistance resulting in lower ligand leakage
  • Generic elution profile for different monoclonal antibodies enables platform approach to purification
  • High-flow agarose matrix allows the processing of large feed volumes
  • Used in the commercial manufacture of monoclonal antibodies

TRUSTED_SUSTAINABILITY

Specifications

Quantity 200 mL
Stability Stable in aqueous Buffers commonly used in Protein A Chromatography
For Use With (Application) For Use of Rigorous and Cost-Effective CIP and Sanitization Protocols, Manufacture of Monoclonal Antibodies
Storage Requirements 2°C to 8°C
Binding Capacity 30 mg Human lgG/mL (at 2.4 min. residence time)
Product Type MabSelect SuRe Media
Flow Rate <500 cm/Hoμr
Particle Size 85 μm
Height (English) Bed 7.87 in.
Height (Metric) Bed 20 cm
Includes Certificate of Analysis
Max. Pressure <200 kPa
Matrix Highly Cross-Linked Agarose
pH Range 3 to 12
Storage Buffer 20% Ethanol
Ligand Type Alkali-stabilized Protein A-derived (E. coli)
Show More Show Less
compliance-icons
Product Identifier
  • MabSelect SuRe, 200 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.