missing translation for 'onlineSavingsMsg'
Learn More

Cytiva MabSelect Sure™ LX Chromatography Media

Product Code. 11693074 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 mL
200 mL
Unit Size:
200mL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11693074 200 mL 200mL
11683074 25 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11693074 Supplier Cytiva Supplier No. 17547402
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protein A affinity medium with high dynamic binding capacity which improves performance and cuts costs

Alkali tolerance allows for stability, thorough cleaning and reduction in contamination

  • Alkali-stabilized Protein A-derived (E.coli) Ligand
  • High dynamic binding capacity (DBC) for high titer cultures
  • Reduces risk of contamination and carryover
  • Alkali-tolerant rProtein A ligand withstands rigorous CIP and sanitization procedures with 0.1 to 0.5 M NaOH

Purification of monoclonal antibodies for clinical applications and process scale

Certifications

Certificate of Analysis

TRUSTED_SUSTAINABILITY

Specifications

Quantity 200 mL
Stability Stable in aqueous Buffers commonly used in Protein A Chromatography
For Use With (Application) For PurifiCatIon of Monoclonal Antibodies for Clinical AppliCatIons
Storage Requirements 2°C to 8°C
Binding Capacity 60 mg Human IgG/mL mediμm
Product Type MabSelect SuRe LX Media
Flow Rate 500 cm/Hoμr
Particle Size 85 μm
Height (English) Bed 7.87 in.
Height (Metric) Bed 20 cm
Includes Certificate of Analysis
Max. Pressure <2 bar
Matrix Highly Cross-Linked Agarose
pH Range 3 to 12
Storage Buffer 20% Ethanol
Ligand Type Alkali-stabilized Protein A-derived (E. coli)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.