missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MacConkey Agar No. 2 (Dehydrated)

Product Code. 10269712
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10269712

Brand: Thermo Scientific™ CM0109B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Detect and enumerate enterococci from water, sewage, food products with this medium.

Detect and enumerate enterococci from water, sewage, food products with Thermo Scientific™ Oxoid™ MacConkey Agar No. 2 (Dehydrated). MacConkey Agar No.2 is a modification of the original MacConkey solid medium and is especially useful for the recognition of enterococci, in the presence of coliforms and non-lactose fermenters from water, sewage, food products, etc.

Use MacConkey Agar No. 2 for detection of coliform organisms in water, wastewater, and dairy products for the presence of indicator groups such as coliforms. Besides safety, indicator groups provide a measure of food quality and spoilage potential. One of the most common groups of bacteria employed as indicator organisms by the food industry are the coliforms, which can be regarded as a subgroup within the Enterobacteriaceae.

  • Easy-to-read: enterococci appear as small intensely red colonies with a pale periphery while non-lactose fermenters appear colorless.
  • Selective: selective for enterococci, while completely inhibiting bile tolerant Gram-positive cocci, such as staphylococci, and non-fecal streptococci.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description MacConkey Agar No. 2 (contains Bile Salts No. 2)
Form Powder
Product Type Agar
Quantity 500 g
Yield For 9.7L medium

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.