missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MacConkey Broth Purple (Dehydrated)

Product Code. 10341433
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10341433 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10341433 Supplier Thermo Scientific™ Supplier No. CM0505B
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detect coliform organisms in water and milk examination with this medium.

Detect coliform organisms in water and milk examination with Thermo Scientific™ Oxoid™ MacConkey Broth Purple (Dehydrated). Bromocresol purple indicator in the medium provides more sensitive and definite indications of acid production as compared to neutral red indicator and is less inhibitory to Escherichia coli. The use of bromocresol purple corresponds to the alternative formulations recommended in The Bacteriological Examination of Water Supplies1 and International Standard for Drinking Water2.

MacConkey Broth has long been used as a presumptive medium for the detection of coli-aerogenes organisms. In the original medium, litmus was employed as the indicator of acid production but, in later publications, MacConkey suggested neutral red as a more satisfactory alternative. Childs & Allen1 showed that some samples of neutral red exerted an inhibitory effect on the growth of Escherichia coli in this medium.

Bromocresol purple is less inhibitory, and the color change from purple to yellow provides a more sensitive and definite indication of acid formation; therefore this indicator is used in MacConkey Broth Purple.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description MacConkey Broth Purple (U.S. formulation, CM5a)
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 12.5L medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.