Learn More
Emma Safety Footwear Mack M Safety Shoes
Product Code. 15908681
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15908681
Brand: Emma Safety Footwear MM53486338
Only null left
Add to basket
Add to basket
A superb shoe for galvanizing and welding metal. A closed front flap protects against stray sparks and hot metal splinters and offers enhanced protection for the metatarsal. Includes polyurethane outer toe protection and a concealed steel toe cap.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • GripForce™ Duo Heat Resistant Outsole
  • Heat and Cold insulated
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Laces
Color Black
Design Mack M
Liner Material Hydro-Tec™ Sanitized Silver
Material Leather
Size 38
Toe Material Steel