missing translation for 'onlineSavingsMsg'
Learn More

Hellma Macro Cuvette, 6030-OG

Product Code. 15810332
Change view
Click to view available options
Path Length:
10 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Path Length Unit Size
15810332 10 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15810332 Supplier Hellma Supplier No. 60301010
  • ONLINE EXCLUSIVE PRICE
    64.10 €
    51.49 € / Pack of 1
    Save 12.61 €
    19% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Optical glass absorption assay cuvette with a PTFE lid.

  • Optical glass construction
  • VIS range
  • PTFE lid included

Specifications

Capacity (Metric) 3500 μL
Material Optical Glass
Closure Material PTFE
Width (Metric) Interior 9.5 mm
Thickness (Metric) Base Thickness: 1.5 mm
Includes PTFE Lid
Quantity 1/Pk
Path Length 10 mm
Wavelength Range 360 to 2500 nm
Height (Metric) 45 mm
Width (Metric) 12.5 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.