Learn More
Berthold Technologies Macro Lens 50 mm F0.95 Ikon Version
Product Code. 31777437 Shop All Berthold Technologies Products
Change view
Click to view available options
Width (Metric):
50 mm
Unit Size:
Each
Product Code. Width (Metric) Unit Size
31777437 50 mm Each
1 options
Product Code. 31777437 Supplier Berthold Technologies Supplier No. 60223
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Used for closeup imaging in the NightSHADE in vivo imaging system.
  • Required when using the Macro table with the NightSHADE system
  • Enables image acquisition at higher magnification
Specifications
Type Macro Lens
For Use With (Equipment) Imaging System
Width (Metric) 50 mm