Learn More
uvex MACSOLE™ Boots S7
Product Code. 30533590 Shop All Uvex Products
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
Unit Size:
1 pair
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
30533590 36 1 pair
30533583 37 1 pair
30533586 38 1 pair
30533582 39 1 pair
30533588 40 1 pair
30533597 41 1 pair
30533602 42 1 pair
30533572 43 1 pair
30533604 44 1 pair
30533569 45 1 pair
30533573 46 1 pair
30533587 47 1 pair
30533601 48 1 pair
30533595 49 1 pair
30533603 50 1 pair
30533584 51 1 pair
30533575 52 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Product Code. 30533590 Supplier uvex Supplier No. 6863336
  • ONLINE EXCLUSIVE PRICE
    181.00 €
    178.27 € / 1 pairs
    Save 2.73 €
    1% Off
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
These uvex 3 MACSOLE™ boots are designed for heavy-duty applications.
The uvex 3, but even more robust.
  • The MACSOLE™ S7 68633 boots with rubber outsole provides even stronger support for heavy-duty applications, making it particularly resistant to heat, chemicals and cuts.
  • The MACSOLE™ rubber sole technology also impresses with its unbeatable slip resistance (SR), even in the cold.
  • EN ISO 20345: S7
TRUSTED_SUSTAINABILITY
Specifications
Color Black, Blue
Material Rubber, Leather
Size 36
Toe Material Polymer
Disposable Reusable
Gender Men's
Fit Wide Width
Latex Free No
Waterproof Yes
Product Type Boots