missing translation for 'onlineSavingsMsg'
Learn More

MP BIOMEDICALS MagBeads FastDNA Kit Ready-to-Use for MPure-32 96 Preps

Product Code. 30067474 Shop All MP Biomedicals Products
Change view
Click to view available options
No. of Samples:
96 Preps
Unit Size:
96 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Samples Unit Size
30067474 96 Preps 96 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30067474 Supplier MP BIOMEDICALS Supplier No. 117033600
  • ONLINE EXCLUSIVE PRICE
    1565.00 €
    1483.65 € / 96 preps
    Save 81.35 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MagBeads FastDNA™ Kit is intended for rapid extraction of DNA from tissue, cells, blood, saliva, swabs, blood spots, semen, and other clinical samples using the MPure-32™ aNAP System.

MagBeads FastDNA™ Kit is based on the purification method of high binding magnetic particles.

  • The sample is lysed and digested.
  • DNA is released into the lysate.
  • After addition of magnetic particles and binding solution, DNA will be adsorbed on the surface of magnetic particles, and impurities such as proteins will be removed without adsorption.
  • The adsorbed particles were washed with washing buffer to remove the proteins and impurities, washed with ethanol to remove salts, and finally the DNA was eluted with Elution Buffer.

Specifications

Format Kit
Isolation Technology Magnetic Bead
Sample Type Tissue, Cells, Blood, Saliva, Swabs, Semen, Other Clinical Samples
For Use With (Application) DNA Extraction
No. of Samples 96 Preps
Product Type DNA Kit
compliance-icons compliance-icons compliance-icons
Product Identifier
  • (MPure-32) MagBeads FastDNA Kit
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sodium Dodecyl Sulphate, n-Hexadecane, Proteinase K, maleicacid, Ethanol, Guanidine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.