Learn More
Melford Magenta-Gal
Product Code. 30018110
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018110 100 mg 100mg
30018111 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30018110 Supplier Melford Supplier No. B730000.1
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
Chromogenic substrate of β-Galactosidase.
Chromogenic substrate of β-Galactosidase. Produce magenta color.
Specifications
Quantity 100 mg