Learn More
Melford Magenta-GlcA
Product Code. p-200086997
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
Product Code. 30018112
Brand: Melford B730500.1
Chromogenic substrate for B-D-Glucuronidase (GUS).
Chromogenic substrate for B-D-Glucuronidase (GUS). Produces a magenta color for (GUS) gene detection.
Specifications
Quantity 100 mg