Learn More
Invitrogen™ MagicMedia™ E. coli Expression Medium, Powder
Beschreibung
Includes
Contains a proprietary Component A (in foil pouch powder format), and Component B (in buffer liquid format).
• Achieve 3–10 times higher protein yields (Figure 1) with much less hands-on time
• Eliminate time-consuming OD monitoring steps—simply inoculate, grow the culture overnight, and harvest (Figure 2)
• Reduce culture volume to simplify handling and scale-up processes
• Ideal for simultaneous expression of multiple clones for high-throughput applications
MagicMedia E. coli Expression Medium, powder, consists of a proprietary Component A (in foil pouch powder format) and Component B (in buffer liquid format).
MagicMedia E. coli Expression Medium is also available in a ready-to-use, pre-sterilized liquid format (Cat. No. K6803).
Bestellinformation
Store Component A (foil pouches) and Component B buffer (liquid) at Room Temperature (15 - 30°C).
Spezifikation
Spezifikation
| Content And Storage | Store Component A (foil pouches) and Component B (liquid) at room temperature (15–30°C). |
| Form | Powder, Liquid |
| Format | Kit |
| Product Type | Microbiology Media |
| Product Line | MagicMedia |
| Quantity | 1 L |
| Species | Escherichia coli |
For Research Use Only. Not for use in diagnostic procedures.
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.