missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™-96 AI/ND Viral RNA Isolation Kit

Product Code. 30054625 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
384 preps
Unit Size:
Pack of 384
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30054625 384 preps Pack of 384
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30054625 Supplier Applied Biosystems™ Supplier No. AM1835
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Applied Biosystems™ MagMAX™-96 AI/ND Viral RNA Isolation Kit is for the isolation of viral RNA and DNA from diverse sample types (i.e., environmental samples and biofluids). Nucleic acids purified using this kit are ideal for PCR and RT-PCR. applications.

The MagMAX™-96 AI/ND Viral RNA Isolation Kit is a first generation MagMAX™ Viral RNA Isolation Kit validated by the NVSL for use with AI/ND viruses. Perfect for working with low viral concentrations, the kit contains sufficient reagents for 4 x 96 reactions.

  • Validated by the NVSL for use with AI/ND viruses
  • Recover as few as 10 copies of viral RNA from 100 to 400 μL sample
  • Typical viral RNA recovery of >50%
  • Ideal when working with low viral concentrations
  • Linear recovery down to 50 transcripts
  • 96 samples can be processed in 30 minutes
  • Easily integrated into robotic platforms

Benefits of MagMAX™ Magnetic Bead-Based Purification:

Magnetic beads offer many benefits compared to other technologies for isolating RNA. Beads bind RNA more efficiently than glass fiber filters, resulting in higher and more consistent RNA yields. Additionally, because filters are not used, there is no risk of filter clogging due to cellular particulates in samples. Since only a small volume of magnetic beads is needed for high-efficiency binding, the bound RNA can be eluted in just 20 to 50 μL of nuclease-free water, concentrating RNA from large, dilute samples.

Optimized for Isolation of Virus from Biofluids:

The Applied Biosystems™ MagMAX™ Viral RNA Isolation Kit has been optimized for use with biological fluids and cell-free samples such as serum, plasma, swabs, and cell culture media. These samples often present unique challenges when purifying RNA, including low viral titers, large volumes to be processed, and sample complexity. Without the need for organic solvents or RNA precipitation, the MagMAX™ Viral RNA isolation kit eliminates the problems often encountered with other RNA isolation methods such filter clogging, large elution volumes, and inconsistent yields.

Get the Same Amount of RNA from 10-fold Less Sample

The sensitivity of viral RNA detection was compared using RNA isolated with Applied Biosystems™ MagMAX™ protocol or the current National Veterinary Services Laboratories (NVSL) validated viral testing protocol. Even at one-tenth the sample input, viral RNA isolated using the MagMAX™ kit could be detected at equal or greater levels than RNA isolated using the NVSL protocol and recommended competitor's kit.

Accessory Products:

Carrier RNA (SKU#4382878), Lysis/Binding Solution Concentrate (SKU#AM8500), Wash Solution 1 Concentrate (SKU#AM8504), and Wash Solution 2 Concentrate (SKU#AM8640) are components of the MagMAX™ Viral RNA Isolation Kits that may also be purchased separately. A 96-well magnetic ring stand is required to use this kit. The Magnetic Stand-96 (SKU#AM10027) and the 96-well Magnetic Ring Stand (SKU#AM10050) are designed for paramagnetic bead precipitation from standard 96 well U-bottom microplates. The Magnetic-Ring Stand pellets the magnetic beads in a circle shape, enabling easy removal of supernatant by a robot or multichannel pipettors. For additional processing plates, Matrix 96-Well U Bottom Polypropylene Plates (Thermo Scientific, Item# 4917) are recommended. The MagMAX™ and MagMAX™-96 Viral RNA Isolation Kits (SKU#AM1939 and SKU#AM1836) are the newest generation MagMAX™ kits for this application, but are not yet validated by the NVSL.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Carrier RNA is stored at –20°C; RNA Binding Beads are stored at 4°C. Wash Solution 2 Concentrate and Elution Buffer may be stored at either 4°C or room temperature. Lysis⁄Binding Solution Concentrate, Wash Solution 1 Concentrate, RNA Rebinding Concentrate, Bead Resuspension Solution, Processing Plates and Lids, and PBS Buffer are stored at room temperature. Nuclease-free water may be stored at any temperature.
Format 96-well Plate
Isolation Technology Magnetic Bead
Purification Time 1 hr.
Sample Type Liquid Samples (e.g. Serum), Viral Samples
Final Product Type Viral RNA
High-throughput Compatibility High-throughput Compatible, Not High-throughput Compatible (Manual), Automated Protocols
For Use With (Equipment) KingFisher™ Flex Magnetic Particle Processor, KingFisher 96, MagMAX™ Express-96 Standard Magnetic Particle Processor
For Use With (Application) Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Northern Blotting
No. of Reactions 384 preps
Product Line MagMAX
Quantity 384 preps
Starting Material Amount Up to 50μl of sample
Target Viral DNA/RNA
Yield 5 ng
Show More Show Less

For Veterinary Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.