Learn More
Description
The Applied Biosystems MagMAX CORE disruption modules are a sample preparation system designed to complement the MagMAX CORE Nucleic Acid Purification Kit. This modular technology is optimized for the purification of nucleic acids from a wide range of sample matrices commonly submitted for veterinary diagnostic testing. The MagMAX CORE disruption modules aid in the processing of hard-to-lyse organisms like Mycobacterium avium paratuberculosis (MAP).
Key features
- Convenience—a complete solution using CORE chemistry
- Confidence—optimized extraction of nucleic acids from difficult-to-lyse samples
- Flexible—purchase only what you need to lyse MAP and other hard-to-lyse organisms; 100 reaction tube module for lower throughput or 500 reaction plate module for higher throughput
- Ergonomy—streamlined sample preparation protocol versus the previous mechanical lysis module
- Sustainability—reduced tube consumption with use of high-throughput plate module
There are two options of module depending on workflow and throughput needs:
- MagMAX CORE Disruption Tubes Module, 100 reactions, contains bead beating tubes prefilled with zirconia beads for effective disruption of individual samples
- MagMAX CORE Disruption Plates Module, 500 reactions, contains 96-deep-well plates prefilled with zirconia beads and foil seal adapted for effective disruption of a higher number of samples
Specifications
Specifications
| Detection Method | qPCR |
| Format | Tube |
| Isolation Technology | Mechanical Disruption |
| Scale | Maxi |
| For Use With (Application) | DNA Purification |
| Quantity | 100 Reactions |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.