missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ CORE Nucleic Acid Purification Kit

Product Code. 30054479 Shop All Applied Biosystems Products
Change view
Click to view available options
No. of Reactions:
100
500
Unit Size:
100 reactions
500 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
30054479 100 100 reactions
30057770 500 500 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30054479 Supplier Applied Biosystems™ Supplier No. A32700
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The MagMAX CORE Nucleic Acid Purification Kit is a highly-effective solution for purifying RNA and DNA from an expansive range of veterinary sample types. The universal chemistry is optimized for use on a variety of KingFisher magnetic particle processors and the kit's modular design enables flexibility to evolve with future testing needs.

The MagMAX CORE kit helps to streamline laboratory workflows for downstream real-time PCR, RT-PCR, and sequencing applications

The MagMAX CORE Nucleic Acid Purification Kit offers:

  • Pre-mixed, room-temperature reagents, greatly improving product handling and storage
  • Minimal extraction and reagent preparation time, to get diagnostic results more quickly
  • Complete workflows that address 14 unique sample types, providing a universal sample preparation solution
  • A single protocol adapted on multiple KingFisher magnetic particle processors, unifying all workflows regardless of throughput

Accelerate

The MagMAX CORE Nucleic Acid Purification Kit provides faster instrument processing times, allowing for diagnostic results more quickly. The kit needs less preparation time, due to pre-mixed washes that do not require the addition of alcohols. The MagMAX CORE workflow provides higher-quality nucleic acids than other on-market chemistries, which leads to fewer workflow interruptions and greater diagnostic sensitivity.

Optimize

The MagMAX CORE Nucleic Acid Purification Kit requires less KingFisher plastics, eliminating unnecessary waste and costs. All kit reagents are stored at room temperature, greatly improving product handling and saving on storage space and costs. The MagMAX kit operates using a single protocol adapted on multiple KingFisher magnetic particle processors, unifing workflows regardless of throughput. The chemistry is optimized to address 15 unique sample types, including:

  • Whole blood
  • Feces
  • Oral fluid
  • Animal, fecal, and environmental swabs
  • Tissue
  • Milk
  • Hair follicles
  • Semen
  • Serum
  • Ear notches
  • Ear punches
  • Environmental samples
  • BioMed Diagnostics InPouch TF (Tritrichomonas foetus) culture
  • Plasma

Evolve

The MagMAX CORE Nucleic Acid Purification Kit is designed to be the last sample preparation you may ever need to validate. It utilizes a 'CORE' chemical formulation and couples easily to flexible modules, introducing new applications without introducing new chemistries. The novel MagMAX CORE kit formulation and workflow prepares samples for downstream sequencing applications.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 1 x 50 mL MagMAX CORE Lysis Solution
• 1 x 45 mL MagMAX CORE Binding Solution
• 1 x 60 mL MagMAX CORE Wash Solution 1
• 1 x 60 mL MagMAX CORE Wash Solution 2
• 1 x 12 mL MagMAX CORE Elution Buffer
• 1 x 2.2 mL MagMAX CORE Magnetic Beads
• 1 x 1.25 mL MagMAX CORE Proteinase K

Store at room temperature.
Sample Type Bacteria, Blood, DNA, Liquid Samples (e.g. Serum), Other Samples, RNA, Tissue, Viral Samples
For Use With (Application) Real-Time PCR, Sequencing
High-throughput Compatibility High-throughput Compatible
No. of Reactions 100
Product Line MagMAX
Product Type Nucleic Acid Purification Kit
Quantity 100 Reactions
Shipping Condition Room Temperature
Target Total Nucleic Acids
When using the MagMAX CORE Nucleic Acid Purification Kit (Cat. No. A32700), I am getting good DNA / RNA concentrations and also acceptable 260/280 ratios, but, however, very low 260/230 ratios (<1), how could I improve this value to be closer to 1.8?

Performing an additional alcohol wash step will help improve this ratio.

With the MagMAX CORE Nucleic Acid Purification Kit, do I need to add alcohol to any of the wash buffers?

No. All buffers arrive pre-mixed with alcohol.

Where can I find the .bdz files for use with the MagMAX CORE Nucleic Acid Purification Kit?

You can find the .bdz files located within the Documents section of the MagMAX CORE product page (https://www.thermofisher.com/order/catalog/product/A32700) under "Product Literature".

What is different about the MagMAX CORE Nucleic Acid Purification Kit compared to the other MagMAX kits?

The MagMAX CORE Nucleic Acid Purification Kit offers pre-mixed, room-temperature reagents, which greatly improve product handling and storage. Specifically, the CORE kit decreases processing times by having pre-mixed washes that do not require the addition of alcohol, a single protocol adapted on multiple KingFisher magnetic particle processors, and a complete protocol that addresses 14 unique sample types.

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.