missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ DNA Multi-Sample Ultra 2.0 Binding Beads

Product Code. 15919130 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
45 mL
Unit Size:
45mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15919130 45 mL 45mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15919130 Supplier Applied Biosystems™ Supplier No. A36579

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

These Magnetic Binding beads are included in the MagMAX DNA Multi-Sample Ultra 2.0 Kit.

These Magnetic Binding beads are included in the MagMAX DNA Multi-Sample Ultra 2.0 Kit. They are available separately for applications that require more binding beads than are provided in the kit.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Product Line MagMAX
Product Type DNA Multi-Sample Ultra 2.0 Binding Bead
Quantity 45 mL
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.