missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ DNA Multi-Sample Ultra 2.0 Wash I Solution

Product Code. 15908850 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1.1 L
Unit Size:
1 pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15908850 1.1 L 1 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15908850 Supplier Applied Biosystems™ Supplier No. A36580
Focus on Regenerative Therapies
Use promo code 28027
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Wash I solution is included in the MagMAX DNA Multi-Sample Ultra 2.0 Kit.

This Wash I solution is included in the MagMAX DNA Multi-Sample Ultra 2.0 Kit. It is available separately for applications that require more Wash Isolution than is provided in the kit.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Product Line MagMAX
Product Type DNA Multi-Sample Ultra 2.0 Wash I Solution
Quantity 1.1 L
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.