missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ DNA Ultra 2.0 with Cell and Tissue Extraction Buffer

Product Code. 16648677
Change view
Click to view available options
Quantity:
1 each
Unit Size:
100 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16648677 1 each 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 16648677 Supplier Applied Biosystems™ Supplier No. A45721

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

This version of the MagMAX DNA Multi-Sample Ultra 2.0 Kit contains an additional buffer that allows you to extract DNA from tissues and cells as well as blood, saliva, buffy coat, and buccal swabs.

The resulting purified gDNA is ideal for a variety of downstream molecular biology applications such as qPCR, next-generation sequencing, and microarray analysis. One kit is sufficient to process 200 x 50–200 μL samples or 100 x 200–400 μL samples.

This version of the MagMAX DNA Multi-Sample Ultra 2.0 Kit contains an additional buffer that allows you to extract DNA from tissues and cells as well as blood, saliva, buffy coat, and buccal swabs.

Key features of the MagMAX DNA Multi-Sample Ultra 2.0 Kit:
• Process 6–96 samples in 45 minutes with 5 minutes hands-on time
• Isolate gDNA from 50 μL–2 mL of sample using a simple protocol in SBS microtiter plates
• Skip sample and buffer volume normalization steps for hassle-free processing
• Download validated BindIt software automation scripts for KingFisher Duo Prime and Flex purification systems
• Automation-friendly stand-alone MagMAX DNA Multi-Sample Ultra 2.0 reagent bottle options available

Isolate genomic DNA from multiple sample types with a single sample prep kit
The MagMAX DNA Multi-Sample Ultra 2.0 Kit isolates gDNA from 50 μL–2 mL of whole blood (figure below), fresh and stabilized saliva (figure below), and EDTA/Streck cfDNA tube buffy coat (figure below) samples. Additionally, the same kit can be used to isolate gDNA from buccal swabs and other biological sample types. The MagMAX DNA Multi-Sample Ultra 2.0 Kit maximizes gDNA yields by treating samples with proteinase K (PK) and PK-enhancer followed by an innovative lysis/binding solution to simultaneously lyse cells, denature proteins, and capture gDNA on the high-binding capacity Ultra 2.0 gDNA magnetic binding beads. The Ultra 2.0 Wash I solution removes proteins, detergents, and other impurities from the bound nucleic acid. The gDNA is then released from the beads using a low-TE elution solution.

Fast and hassle-free MagMAX magnetic bead-based gDNA workflows
The MagMAX DNA Multi-Sample Ultra 2.0 Kit combines multiple laborious sample processing steps together into a single step to minimize the time and effort traditionally required by other methods without compromising yield and purity. Process a broad range of sample volume inputs simultaneously without sample normalization or reagent volume adjustments for hassle-free sample prep. Simply match your sample volumes to one of three standard input workflows (50 μL to 400 μL, up to 1 mL, up to 2 mL) and follow the work instructions for either manual or semi-automated sample preparation.

Made for KingFisher purification systems
The MagMAX DNA Multi-Sample Ultra 2.0 Kit is optimized for use on the moderate-throughput KingFisher Duo Prime (Cat. No. 5400110) and the higher-throughput KingFisher Flex (Cat. No. 5400630) purification systems. Users can process up to 400 μL of sample volume in high-throughput SBS 96-well deep-well microtiter plates using the Flex system. For smaller sample batches, users can process 12 samples using the Duo Prime system. Larger sample input volumes (1–2 mL), which are more challenging to process, are purified in SBS 24-well deep-well microtiter plates for either 24 samples on the Flex or 6 samples on the Duo Prime system. KingFisher instrument BindIt software scripts for all sample types and both Flex and Duo Prime systems can be downloaded below and installed onto the KingFisher instruments.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 1 MagMAX DNA Multi-Sample 2.0 Kit
• 60 mL Cell and Tissue DNA Extraction Buffer
Sample Type Cells, Tissue
For Use With (Application) Next-Generation Sequencing, PCR, Real-Time PCR
High-throughput Compatibility Automated Protocols, High-throughput Compatible, Manual Protocols
Product Line MagMAX
Product Type DNA Multi-Sample Ultra Kit
Quantity 1 each
Shipping Condition Room Temperature
Target Genomic DNA

For research use only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.