missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ mirVana™ RNA Isolation Kits

Product Code. 15422143 Shop All Applied Biosystems Products
Change view
Click to view available options
Product Type:
Kit
Quantity:
96 reactions
Unit Size:
96 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Quantity Unit Size
15422143 Kit 96 reactions 96 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 15422143 Supplier Applied Biosystems™ Supplier No. A27828
Keep Your Science Moving with 3 for 2
Use promo code 27970
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price

The MagMAX™ mirVana™ Total RNA Isolation Kit is designed for isolation of total RNA, including small RNAs such as microRNAs, from a wide variety of sample types.

Scalable isolation of RNA (including small RNAs)

The MagMAX™ mirVana™ Total RNA Isolation Kit is designed for isolation of total RNA, including small RNAs such as microRNAs, from a wide variety of sample types. The kit uses MagMAX magnetic-bead technology, enabling reproducible recovery of high-quality RNA that is suitable for a broad range of applications, including TaqMan™ miRNA Detection Assays.

The magnetic bead–based purification format allows you to easily scale from processing 6 to 96 samples at once when used with the KingFisher™ Duo Prime or KingFisher™ Flex Magnetic Particle Processor. Alternatively, samples can be processed manually with the use of a magnetic stand.

Features of the MagMAX mirVana Total RNA Isolation Kit include:

  • Automation-ready, phenol-free extraction
  • Streamlined protocols for numerous non-invasive biological samples as well as tissues and cell culture
  • Recovery of pure miRNAs compatible with miRNA-Seq and qRT-PCR methods such as the TaqMan™ Advanced miRNA cDNA Synthesis Kit and TaqMan™ Advanced miRNA Assays

One kit to isolate true total RNA from a diverse set of sample types

Why purchase multiple purification kits to isolate all RNAs present in your different sample types? The MagMAX mirVana Total RNA Isolation Kit eliminates the headache of ordering 'specialized' kits because it has been optimized for use with most biological samples. No longer are separate, specific kits needed to isolate microRNAs from plasma and solid tissue samples.

The kit is designed for both low- and high-throughput isolation of true total RNA that covers the full range of RNAs present in cells from small RNAs such as microRNAs to non-coding RNA, messenger RNA, and ribosomal RNA. The phenol-free, magnetic-bead format makes the kit fully amenable to automated RNA isolation from the following samples:

  • Serum/plasma
  • Whole blood
  • Tissue
  • Cell culture
  • Urine

Benefits of MagMAX magnetic bead–based purification

Magnetic beads offer many benefits compared to other technologies for isolating total RNA. Beads bind RNA more efficiently than glass fiber filters, thereby enabling higher and more consistent yields. Additionally, because filters are not used, there is no risk of filter clogging due to cellular particulates in samples.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

Box 1 (store at -15 to -25°C)
• 1 x 0.48 mL Proteinase K, 20 mg/mL
• 1 x 0.96 mL Lysis/Binding Enhancer
• 1 x 0.2 mL TURBO DNase, 20 U/uL

Box 2 (store at room temperature)
• 1 x 115 mL Lysis Buffer
• 1 x 4.4 mL PK Digestion Buffer
• 1 x 2 mL RNA Binding Beads
• 1 x 20 mL Wash Solution 1 Concentrate
• 1 x 60 mL Wash Solution 2 Concentrate
• 1 x 4.8 mL Rebinding Buffer
• 1 x 4.8 mL MagMAX TURBO DNase Buffer
• 1 x 9.6 mL Elution Buffer
• 1 Processing Plate
• 2 Elution Plates
• 4 Plate Covers

Isolation Technology Magnetic Bead
Purification Time Up to 1.5 hr.
Elution Volume 20 to 100 μL
Sample Type Cells, Plasma, Serum, Tissue, Urine, Whole Blood
Final Product Type Total RNA
High-throughput Compatibility High-throughput Compatible
For Use With (Equipment) KingFisher™ Duo Prime, KingFisher™ Flex, KingFisher™ Apex
For Use With (Application) RT-PCR, qPCR, cDNA library construction, NGS, microarray analysis, blot hybridization, Northern blotting, in vitro translation, nuclease protection assays, nucleic acid labeling
Product Type Kit
Quantity 96 reactions
Shipping Condition Box 1: Dry Ice, Box 2: Room Temperature
Starting Material Amount Cells: ≤106, Plasma: ≤100 μL, Serum: ≤100 μL, Tissue: ≤50 mg, Urine: ≤250 μL, Whole Blood: ≤50 μL
Target Total RNA, mRNA, siRNA, miRNA
Yield Cells: ≤32 μg RNA per 50,000 cells, Tissue: ≤500 μg per 5 mg
Show More Show Less
Can I purchase the RNA Binding beads from the MagMAX MirVana Total RNA Isolation Kit (Cat. No. A27828) separately?

Yes, you can. The DNA Binding Beads for MagMAX-96 DNA Multi-Sample Kit (Cat. No. 4489112) are the same as that used in A27828.

What is the minimum cell number for RNA isolation using MagMAX mirVana Total RNA Isolation Kit (Cat. No. A27828)?

We have tested extraction from as low as 1,000 cells using cultured HeLa, Jurkat, and HepG2 cells.

What is the level of detection for somatic variants with the Oncomine Myeloid Research Assay GX?

Detection of somatic variants with the Oncomine Myeloid Research Assay GX has been verified down to 5% allele frequency.

Which extraction kits are compatible with Oncomine Myeloid Research Assay GX?

Here is a list of kits that are compatible with Oncomine Myeloid Research Assay GX:

Nucleic acid isolation - RNA samples
- MagMAX mirVana Total RNA Isolation Kit, Cat. No. A27828
- PureLink Total RNA Blood Kit, Cat. No. K156001
- RNaseZap RNase Decontamination Solution, Cat. No. AM9780, AM9782, AM9784

Nucleic acid isolation - DNA samples
- MagMAX DNA Multi-Sample Ultra 2.0 Kit, Cat. No. A36570
- PureLink Genomic DNA Mini Kit, Cat. No. K1820-01

What are the sample input requirements for Oncomine Myeloid Research Assay GX?

The recommended minimum sample input is 27.75 ng of DNA (1.11 ng/µL minimum concentration) and 14.25 ng of RNA (0.95 ng/µL minimum concentration).

What sample controls should I use with the Oncomine Myeloid Research Assay GX?

The recommended controls are:

DNA Controls:
- Seraseq® Myeloid Mutation DNA Mix, Cat. No. 0710-0408 (SeraCare®)
- Myeloid DNA Reference Standard, Cat. No. HD829 (Horizon™)
- AcroMetrix™ Oncology Hotspot Control, Cat. No. 969056

RNA Controls:
- Universal Human Reference RNA, Cat. No. 740000 (Agilent™)
- Seraseq® Myeloid Fusion RNA Mix, Cat. No. 0710-0407 (SeraCare®)

How many Oncomine Myeloid Research Assay GX samples can be multiplexed on a GX5 Chip?

Multiplex sequencing of up to 8 Oncomine Myeloid Research Assay GX samples (8 DNA+ 8 RNA) per lane on a GX5 Chip can be performed in a single run. There are 4 lanes on the GX5 Chip, so 32 total samples (32 DNA+ 32 RNA) can be run on one GX5 Chip (8 samples per run).

What are the components of Oncomine Myeloid Research Assay GX?

The Oncomine Myeloid Research Assay GX (Cat. No. A47857) includes the 2-pool DNA panel, the 1-pool RNA panel, Genexus Strip 1, and Genexus Strip 2-AS. The DNA panel is provided in two packs of 8 tubes (4 tubes of Myeloid DNA Pool 1 and 4 tubes of Myeloid DNA Pool 2 per pack). The RNA panel is provided in one pack of 8 tubes (Myeloid RNA Pool 1). Each primer pool in the panel is provided in pairs of tubes, where each tube pair contains one tube with primers in position 1 and one empty uncapped tube in position 2. Three 8-strip packs of the Genexus Strip 1 and Genexus Strip 2-AS (24 total of each strip) are provided with each kit.

How many samples can I run using Oncomine Myeloid Research Assay GX?

The contents of each Oncomine Myeloid Research Assay GX kit are sufficient for up to 32 samples (32 x 3‑pool reactions: 32 x 2‑pool DNA reactions and 32 x 1‑pool RNA reactions).

How many primer pools are present in the Oncomine Myeloid Research Assay GX?

The Oncomine Myeloid Research Assay GX includes 3 pools of Ion AmpliSeq oligonucleotide primers (a 2-pool DNA panel and a 1-pool RNA panel).

What is the Oncomine Myeloid Research Assay GX gene content?

The Oncomine Myeloid Research Assay GX targets key genes and fusions associated with major myeloid disorders, including acute myeloid leukemia (AML), myeloid dysplastic syndrome (MDS), myeloproliferative neoplasms (MPN), chronic myeloid leukemia (CML), chronic myelomonocytic leukemia (CMML), and juvenile myelomonocytic leukemia (JMML). The DNA panel comprises 40 key genes while the RNA panel includes a broad fusion panel of 29 driver genes, enabling detection over 600 unique fusion isotypes. See flyer for gene targets (https://assets.thermofisher.com/TFS-Assets/CSD/Flyers/Oncomine_myeloid_assay_gx_flyer.pdf).

What is the Oncomine Myeloid Research Assay GX designed for ?

The Oncomine Myeloid Research Assay GX is a comprehensive targeted next-generation sequencing (NGS) assay designed for sensitive detection of myeloid disorder-associated DNA mutations and fusion transcripts in blood and bone marrow samples.

I am getting MagMAX bead carryover in some wells of my plate. What might be causing this and how can I resolve it?

Some bead carryover is common and has not been documented to affect downstream assays. The most common cause of bead carryover is too much input sample. The best way to determine if the sample amount is the cause of the issue is to run PBS with Xeno (artificial DNA/RNA) in the lysis buffer and then check if there is still bead carryover. If that is not the cause, the initial sample may require special processing, such as bead beating or liquification. If there is still bead carryover without the original sample, and in particular, if the carryover is present in the same wells for multiple runs, then this may be an instrument alignment issue.

Do I need any additional consumables when using MagMAX kits on the MagMAX Express-96 instrument or a KingFisher instrument?

Plastic consumables need to be purchased separately in order to use the MagMAX reagents on the MagMAX Express-96 or a KingFisher instrument. The particular consumables and the amount required will vary by kit and protocol so please check the user guide for the particular MagMAX kit to be sure that all required plastic consumables are available prior to sample processing.

Where can I find Bindlt script protocol (.bdz) files for use of MagMAX kits with the KingFisher Flex Purification System and KingFisher Duo Prime Purification System?

The best place to download KingFisher Flex and KingFisher Duo Prime protocols for a particular MagMAX kit is the relevant MagMAX kit product page. The Bindlt script protocol (.bdz) files can be found under the Documents>Product literature section.

Can I scale up the MagMAX kit reaction volumes? If so, how should I do this?

Yes, reaction volumes for MagMAX kits can be scaled up. In general, the amount of MagMAX reagents should be scaled up proportionally for the increase in sample input amount. Please see the specific kit user guide for any recommendation. If none are included, then optimization may be necessary.

Will addition of BME (2-mercaptoethanol) to the MagMAX lysis buffer cause any issues with nucleic acid extraction?

Addition of BME (2-mercaptoethanol) to the lysis buffer of MagMAX kits is fine. This can be beneficial with more RNase-rich samples like certain types of tissues.
In general, we recommend adding 0.3 µL BME per 100 µL of lysate.

Can I store lysates made using the lysis buffer from the MagMAX mirVana Total RNA Isolation Kit (Cat. No. A27828) at -20 degrees C?

Lysates made using the lysis buffer from the MagMAX mirVana Total RNA Isolation Kit can be stored at -20 to -80 degrees C for up to 4 days if 2-mercaptoethanol or 100 mM DTT is added to the lysis buffer prior to homogenization of the sample.

What recommendations do you have for homogenizing tissue samples for the MagMAX mirVana Total RNA Isolation Kit (Cat. No. A27828)?

For tissues containing low to normal levels of cellular RNase (for example, brain, heart, or liver) we recommend using 20 µL of Lysis Binding Mix for 1 mg of tissue (1:20 ratio). For example, use 200 µL of Lysis Binding Mix for 10 mg of tissue.

For tissues containing high levels of cellular RNase (for example, spleen or pancreas), we recommend using 40 µL of Lysis Binding Mix for 1 mg of tissue (1:40 ratio). For example, use 400 µL of Lysis Binding Mix for 10 mg of tissue.

We recommend homogenizing for 30 s for full lysis. Our protocol recommends use of single sample polytron homogenizer, but you can also use bead-beating instrument with glass or zirconia beads. You may need to optimize bead-beating for your specific sample.

Is the MagMAX mirVana Total RNA Isolation Kit (Cat. No. A27828) compatible with Tempus/PAXgene stabilized blood samples?

No, only certain MagMAX kits are compatible with Tempus/PAXgene stabilized blood samples. We recommend using only MagMAX kits that are designed for blood samples stored in the designated RNA blood tubes for the kit. For PAXgene tubes, the compatible kit is the MagMAX for Stabilized Blood Tubes RNA Isolation Kit, compatible with PAXgene Blood RNA Tubes (Cat. No. 4451894). For Tempus tubes the compatible kit is the MagMAX for Stabilized Blood Tubes RNA Isolation Kit, compatible with Tempus Blood RNA Tubes (Cat. No. 4451893).

Can frozen serum/plasma samples be used with the MagMAX mirVana Total RNA Isolation Kit (Cat. No. A27828)?

Frozen serum/plasma samples can be used with the MagMAX mirVana Total RNA Isolation Kit, though we recommend fresh samples. We also recommend limiting freeze/thaw cycles to 1-2 cycles to prevent RNA degradation.

Can I use frozen blood samples with MagMAX mirVana Total RNA Isolation Kit (Cat. No. A27828)?

Frozen whole blood samples can be used with the MagMAX mirVana Total RNA Isolation Kit if the downstream application can use fragmented RNA (e.g., qRT-PCR, amplicon sequencing). However, collection of blood samples in Tempus Blood RNA Tubes (Cat. No. 4342792) or PAXgene tubes is recommended for long-term storage and when high-quality RNA is needed because freezing or prolonged storage at 4 degrees C (>24 hrs) results in RNA degradation.

Can I process a larger amount of cultured cells with MagMAX mirVana Total RNA Isolation kit (Cat. No. A27828) than the 1x10^6 cells indicated in the user guide?

The MagMAX mirVana Total RNA Isolation Kit user guide recommends up to 1x10^6 cells, but this can potentially be increased to 5x10^6 cells by increasing the lysis and binding reagents accordingly. This may not work well for all cell types, so we recommend testing control samples of the cell line of interest.

Can I scale up the input volume of whole blood to 150-200 µL with the MagMAX mirVana Total RNA Isolation Kit (Cat. No. A27828)?

Yes, the input can be increased up to 150-200 µL for the automated protocol with all binding reagents kept proportionate.

Can I increase the sample amount to 200 µL for the serum/plasma protocol for the MagMAX mirVana Total RNA Isolation Kit (Cat. No. A27828)?

The input for the serum/plasma protocol for the MagMAX mirVana Total RNA Isolation Kit can be increased to 200 µL of sample for the automated protocol if all binding reagents are increase proportionally.

What is the maximum amount of tissue input for the MagMAX mirVana Total RNA Isolation Kit (Cat. No. A27828)?

The maximum input amount ranges from 2.5-10 mg of tissue depending on the tissue type. For example, spleen and thymus use a lower input amount. However, you can homogenize/lyse a larger piece and store the remaining lysate at -20 degrees C.
Further information regarding tissue input amounts can be found in the MagMAX mirVana Total RNA Isolation Kit (Tissue Samples) User Guide: https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0011132_A27828_magmax_rnaisolation_tissues_ug.pdf

Do you have a script for using the MagMAX mirVana Total RNA Isolation Kit (Cat. No. A27828) on the MagMAX Express 96 instrument?

Unfortunately, we do not have a script for using the MagMAX mirVana Total RNA Isolation Kit (Cat. No. A27828) on the MagMAX Express 96 instrument. However, we do have a free BindIt 4.0 software for converting KingFisher Flex scripts to MagMAX Express 96 scripts.

Do you offer the Proteinase K from the MagMAX mirVana Total RNA Isolation Kit (Cat. No. A27828) separately from the kit?

We do not sell the Proteinase K from the MagMAX mirVana Total RNA Isolation Kit as a standalone item. A different Proteinase K product (Cat. No. AM2548) can be purchased as a standalone item and can be used at 2.5X in the MagMAX mirVana protocol if you would like to add more Proteinase K to the procedure.

Can I use the MagMAX mirVana Total RNA Isolation Kit (Cat. No. A27828) to isolate RNA from exosomes?

The MagMAX mirVana Total RNA Isolation Kit can be used to isolate RNA from exosomes. We first recommend isolating/concentrating exosomes with our Total Exosome Isolation Reagent into a volume of 100 µL (bring up with 1X PBS if needed) and then extracting the RNA using the serum/plasma protocol for MagMAX mirVana Total RNA Isolation Kit. The Proteinase K treatment will work well in place of the organic extraction that is normally used to lyse open the exosomes.

Which MagMAX kit do you recommend for 10µL of whole blood samples?

We have not tested any MagMAX kit with less than 50µL of whole blood. It may be possible to isolate RNA from 10µL of whole blood using the MagMAX mirVana Total RNA Isolation Kit (Cat. No. A27828) by bringing the sample volume up to 50 µL using 1X PBS and then processing the sample using the standard protocol.

I am not using the recommended orbital shaker with the MagMAX mirVana Total RNA Isolation Kit (Cat. No. A27828). Can you recommend specific shake speed settings to use?

The following are speed settings that we recommend using in conjunction with the MagMAX mirVana Total RNA Isolation Kit protocol:
- Speed 2 - 400 rpm
- Speed 4 - 700 rpm
- Speed 10 - 1150 rpm
We still recommend watching your samples to ensure that there is no spill over.

Is the TURBO DNase in the MagMAX mirVana Total RNA Isolation Kit (Cat. No. A27828) the same as the standalone TURBO DNase (Cat. No. AM2239)?

The TURBO DNase in the MagMAX mirVana Total RNA Isolation Kit is the same enzyme as the standalone TURBO DNase, but the concentrations of the enzyme differs and therefore it is not an exact equivalent. The TURBO DNase in the MagMAX mirVana Total RNA Isolation Kit is at 20 U/µL, whereas the standalone TURBO DNase is at 2 U/µL.

Where can I find the scripts for automated use of the MagMAX mirVana Total RNA Isolation Kit?

You can find scripts for using MagMAX mirVana Total RNA Isolation Kit with the MagMAX Express-96, KingFisher Flex, or KingFisher Duo instrument, on the product page (http://www.thermofisher.com/order/catalog/product/A27828).

Can I store and reuse the Proteinase K Digestion Mix that I prepared for the MagMAX mirVana Total RNA Isolation kit?

No, we recommend preparing the Proteinase K Digestion Mix fresh each time.

Can I store my lysed samples in Lysis Binding Mix before starting with the MagMAX mirVana Total RNA Isolation protocol?

Yes, you can, but we only recommend storing the lysed samples in Lysis Binding Mix at -20°C for up to 4 days. Samples should be thawed to room temperature before proceeding with the RNA purification.

Does the MagMAX mirVana Total RNA Isolation Kit come with plates?

Yes, it comes with one processing plate, two elution plates, and four plate covers to support manual processing. However, you will have to purchase deep-well plates, standard plates, and tip combs in addition to that if you wish to process on a KingFisher instrument.

Do you offer a MagMAX RNA isolation kit that I can use for extraction of small RNA such as miRNAs?

Yes, the Applied Biosystems MagMAX mirVana Total RNA Isolation Kit (Cat. No. A27828) will isolate both small and large RNAs from a variety of samples (serum, plasma, blood, tissue, cells, urine).

Can I edit the manufactured KingFisher Flex protocols?

No, manufactured protocols are locked for editing, but they can be renamed using the ‘save as' function and edited as a new version.

Can I use magnetic beads from other vendors with the KingFisher Flex instrument?

For nucleic acid extraction, we recommend using our MagMAX kits and MagJET kits. Other magnetic beads should work well as long as they are 0.5 to 10 µm in size. However, you will have to write your own KingFisher protocols using the Thermo Scientific BindIt software and validate/optimize the protocol on your own.

I am working with liquid samples with high volume and want to extract nucleic acid from them using the KingFisher Flex instrument. Can I use more volume than the protocol advises?

Yes, this can be done by sample splitting. Set the protocol so that the magnet will collect samples from multiple wells before continuing to the next step. Be sure not to overload.

Can I scale up my reaction volume with the KingFisher Flex instrument?

Yes, reaction volumes can be scaled up, and all other volumes (i.e. wash volumes) will have to be increased by the same fold difference. It is generally not recommended to scale up reaction volumes more than 2-fold unless the plate's volume capacity will allow it.


compliance-icons compliance-icons compliance-icons
Product Identifier
  • MagMAX mirVana Total RNA Isolation Kit
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 C
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Thiocyanic acid, Proteinase

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.