missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Plant RNA Isolation Kit

Product Code. 15796048
Change view
Click to view available options
Quantity:
1000 Preps
384 Preps
96 Preps
Unit Size:
1 set
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15796048 384 Preps 1 set
15786048 96 Preps 1 set
17153621 1000 Preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15796048 Supplier Applied Biosystems™ Supplier No. A33899
Simplify Your Gene Expression Workflow
Use promo code 27975
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

The MagMAX Plant RNA Isolation Kit is designed for automated high-throughput or manual purification of total RNA from wide variety of plant species and tissue types.

The MagMAX Plant RNA Isolation Kit is designed for automated high-throughput or manual purification of total RNA from wide variety of plant species and tissue types. It utilizes paramagnetic bead technology, enabling high yields and robust performance. The high binding capacity, uniform particle size, and rapid magnetic response of MagMAX magnetic beads makes the technology ideal for high-throughput automatic nucleic acid purification, as well as for manual nucleic acid purification by low- throughput users. The resulting high quality purified RNA is free of proteins, nucleases, and other contaminants or inhibitors, and can be used in a wide range of downstream applications, such as RT-PCR, RT-qPCR, and other enzymatic reactions.

Versatile—suitable for any plant type and part, including woody, polyphenol-rich, and lignified samples
High yields of pure total RNA
High quality—isolates intact total RNA, free of inhibitors and gDNA contamination. Isolated RNA is suitable for long term storage at -20°C.
Flexible—easily adjustable for high-throughput automation platforms and manual processing

Applications
The MagMAX Plant RNA Isolation Kit is designed for fast extraction of high purity RNA suitable for all conventional molecular biology applications, including:
• RT-PCR
• RT-qPCR
• Northern blotting
• Nuclease protection assay

Benefits of MagMAX magnetic bead–based purification
The magnetic bead–based purification format of the kit allows for easy scaling of the number of samples processed either manually with a magnetic stand or with a magnetic particle processor such as the KingFisher Duo Prime or KingFisher Flex processors.

Magnetic beads offer many benefits compared to other technologies for isolating nucleic acids. Beads bind the RNA and DNA more efficiently than glass fiber filters, resulting in higher and more consistent yields. Additionally, because filters and vacuum manifolds are not used, there is no risk of undigested tissue particulates clogging these items during the extraction process. This clogging issue is of particular concern with fibrous plant tissue samples that that are not fully lysed during the disruption and homogenization steps.

Optimized bead surface chemistry
The Dynabeads MyOne Carboxylic Acid utilized in the MagMAX Plant RNA Isolation Kit has an optimized surface chemistry with high specific surface area that allows efficient kinetics and high sensitivity in nucleic acid capture. These aspects make Dynabeads magnetic technology ideal for the recovery of the RNA targets found in a variety of plant samples. Once captured, the RNA is then eluted in 100 μL and ready for downstream applications.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 60 mL lysis buffer, store at room temperature
• 4 vials Dnase I (lyophilized), store at -5 to -30°C after reconsitution
• 4 x 1 mL DNase I reconstitution buffer (to be added to DNase I vial)
• 12 x 1 mL manganese chloride solution, store at -5 to -30°C
• 8 x 1.4 mL RNA binding beads, store at 2°C to 8°C
• 4 x 12 mL DNase I buffer (2X), store at -20° ± 5°C
• 4 x 110 mL Wash Solution 1 concentrate, store at room temperature
• 4 x 50 mL Wash Solution 2 concentrate, store at room temperature
• 4 x 20 mL rebinding buffer, store at room temperature
• 4 x 30 mL elution buffer, store at room temperature

Sample Type Plant Samples
High-throughput Compatibility High-throughput Compatible, Manual Protocols, Automated Protocols
For Use With (Application) Sequencing, RT-PCR (Endpoint), Real-Time PCR
Product Line MagMAX
Quantity 384 Preps
Shipping Condition Room Temperature
Target Total RNA
How should I prepare the MagMAX Plant RNA Kit (Cat. Nos. A33784, A33899, A47157) DNase I solution?

To prepare the MagMAX Plant RNA Kit (Cat. Nos. A33784, A33899) DNase I solution, add 440 µL of DNase I Reconstitution Buffer to each vial of DNase I (lyophilized) and incubate at room temperature for 5 minutes.

To prepare the MagMAX Plant RNA Kit (Cat. No. A47157) DNase I solution, add 10.5 mL of DNase I Reconstitution Buffer to the vial of DNase I (lyophilized) and incubate at room temperature for 5 minutes.

What kind of Polyvinylpyrrolidone (PVP) do you recommend using with MagMAX Plant DNA and RNA isolation kits since there are different kinds available?

For the MagMAX Plant DNA Isolation Kit, we recommend using PVP M.W. 40,000, Cat. No. J62417.A1 (https://www.thermofisher.com/order/catalog/product/J62417.A1).

For the MagMAX Plant RNA Isolation Kit, we recommend using Plant RNA Isolation Aid, Cat. No. AM9690 (https://www.thermofisher.com/order/catalog/product/AM9690).

Can I use the MagMAX Plant RNA Isolation Kit (Cat. No. A33784) to isolate small RNAs?

The MagMAX Plant RNA Isolation Kit has not been optimized for the isolation of small RNAs.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.